Diammonium hydrogen orthophosphate 40% (w/v) in water matrix modifier solution
Supplier: Ricca Chemical
|
Warning
|
Synonyms:
Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
RMMAP40W-50N
RCMMAP40W-50EA
951
USD
RCMMAP40W-50
Diammonium hydrogen orthophosphate 40% (w/v) in water matrix modifier solution
di-Ammonium hydrogen orthophosphate
Formula:
(NH₄)₂HPO₄ Density: 1.2100 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00010891 CAS Number: 7783-28-0 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...