Ammonium nitrate 5% in nitric acid 2% matrix modifier solution

Supplier: Ricca Chemical

RMMAN5N0-50N
RCMMAN5N-50EA 654 USD
RCMMAN5N-50
Ammonium nitrate 5% in nitric acid 2% matrix modifier solution
Ammonium nitrate
Formula: NH₄NO₃
MDL Number: MFCD00011425
CAS Number: 6484-52-2
UN: 1942
ADR: 5.1,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR