Zinc sulfate 0.100 M (M/10) in aqueous solution

Supplier: Ricca Chemical

R9662000-1A
RC9662-32EA 142 USD
RC9662-32
Zinc sulfate 0.100 M (M/10) in aqueous solution
Zinc sulphate
Formula: O₄SZn
MW: 161.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0
EINECS: 231-793-3
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 0.1000 ± 0.0001 M at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR