Zinc sulfate 0.0500 M (M/20) in aqueous solution
Supplier: Ricca Chemical
|
|
R9660000-1A
R9660000-500A
RC966032EA
121
USD
RC966032
RC9660-16
Zinc sulfate 0.0500 M (M/20) in aqueous solution
Zinc sulphate
Formula:
O₄SZn MW: 161.45 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011302 CAS Number: 7733-02-0 EINECS: 231-793-3 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 915) | 0.05000 ± 0.00001 M at 20.0 °C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...