Zinc sulfate 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

R9620000-500A
RC962016EA 81 USD
RC962016
Zinc sulfate 10% (w/v) in aqueous solution
Zinc sulphate
Formula: O₄SZn
MW: 161.45 g/mol
MDL Number: MFCD00011302
CAS Number: 7733-02-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index 10.0 ± 0.6 % (w/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR