Eriochrome Black T 0.5% (w/v) in 2-methoxyethanol for hardness of water determinations

Supplier: Ricca Chemical

R9300000-500C R9300000-120C
RC930016EA 82 USD
RC930016 RC93004
Eriochrome Black T 0.5% (w/v) in 2-methoxyethanol for hardness of water determinations
Eriochrome Black T

In 2-Methoxyethanol.

Formula: C₂₀H₁₂N₃NaO₇S
MDL Number: MFCD00003935
CAS Number: 1787-61-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Dark red, Methoxyethanol odor
Suitability for Use Wine Red (excess Ca or Mg) - Pure Blue (excess EDTA)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR