Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers

Supplier: Ricca Chemical

R9150000-10F R9150000-500A R9150000-4A R9150000-20F R9150000-20E1 R9150000-1A R9150000-4F R9150000-55E
RC915025EA 100 USD
RC915025 RC9150-500 RC91501 RC91505 RC9150-5HP RC9150-32 RCR9150000-4F RCR9150000-55E RCR91500004X4A
Water ACS, Reagent Grade ASTM Type I, ASTM Type II Packaged in plastic containers
Water

This product is manufactured by a continuous process that has been validated to produce high purity Water meeting, as a minimum, the specifications below at the time of packaging. This water also meets ASTM Type III and Type IV specifications.

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Colorless liquid
Chloride (Cl) max 1 ppb
Conductivity at 25°C (Method: SQCP031) max 0.056 µS/cm
Heavy Metals (as Pb) max 0.01 ppm
Nitrate (NO₃) max 0.4 ppm
Organic Carbon (TOC) max 50 ppb
Oxidizable Substances (Permanganate Retention) To Pass Test
Phosphate (PO₄) max 1.0 ppm
Silicate (as SiO₂) max 3 ppb
Sodium (Na) max 1 ppb
Sulfate (SO₄) max 1.0 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR