Barium nitrate USP Test Solution (TS)

Supplier: Ricca Chemical

R0899500-500A
RC899.5-16EA 129 USD
RC899.5-16
Barium nitrate USP Test Solution (TS)
Barium nitrate
Formula: Ba(NO₃)₂
MDL Number: MFCD00003442
CAS Number: 10022-31-8
UN: 1446
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR