Trichloroacetic acid (TCA) 10% (w/v) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

R8685000-500C R8685000-1C R8685000-120C R8685000-4C
RC868516EA 101 USD
RC868516 RC868532 RC8685-4 RCR8685000-4C
Trichloroacetic acid (TCA) 10% (w/v) in aqueous solution analytical reagent
Trichloroacetic acid (TCA)
Formula: Cl₃CCOOH
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54 - 56 °C
Density: Max. 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 2564
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 9.96-10.04 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR