Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA, for Fluoride Analysis using Ion Selective Electrodes, Ricca Chemical Company
Supplier: Ricca Chemical
8670-2.5
RC8670-2.5EA 777 USD
RC8670-2.5
Total Ionic Strength Adjustment Buffer (TISAB II), with CDTA, for Fluoride Analysis using Ion Selective Electrodes, Ricca Chemical Company
Buffers Ionic Strength Adjustment Buffers
Suitable for use in the following compendial methods: -APHA 4500-F C, Reagent 3c -EPA (SW-846) Method 9214, Reagent 5.7 -EPA 340.2, Reagent 6.1