Barium chloride 25% (w/v) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Warning

R0860000-500A
RC860-16EA 152 USD
RC860-16
Barium chloride 25% (w/v) in aqueous solution analytical reagent
Barium chloride
Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: 1560
Melting Pt: 963
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 3287
ADR: 6.1 III
Merck Index: 12,00998

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index 25.0 ± 0.5 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR