Barium chloride 10% (w/v) in aqueous solution

Supplier: Ricca Chemical
Warning

R0850000-4A R0850000-500A R0850000-1A R0850000-20F
RC8501EA 3369 USD
RC8501 RC85016 RC85032 RC850-5
Barium chloride 10% (w/v) in aqueous solution
Barium chloride
Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 3287
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
pH at 25 °C (traceable to NIST SRM 186 & 191) 6.7 ± 0.2 pH at 25.0 °C
Refractive Index 10.0 ± 0.4 % (w/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR