L(+)-Tartaric acid 10% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid

8371-16
RC8371-16EA 63 USD
RC8371-16
L(+)-Tartaric acid 10% (w/v) in aqueous solution
L(+)-Tartaric acid
Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0442 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 10 ± 0.5 % (w/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR