Sulfuric acid 4.00 N in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

R8322000-4A R8322000-1A R8322000-20F R8322000-500A
RC8322-1EA 153 USD
RC8322-1 RC8322-32 RC83225 RCR8322000500A
Sulfuric acid 4.00 N in aqueous solution analytical reagent
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 3.999-4.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR