Sulfuric acid 3.20 N

Supplier: Ricca Chemical

R8321000-4A
RC8321-1EA 829 USD
RC8321-1
Sulfuric acid 3.20 N
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 3.200 ± 0.001 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR