Sulfuric acid 3.00 N in aqueous solution analytical reagent

Supplier: Ricca Chemical
Warning

R8320000-1A R8320000-4A R8320000-20F
RC8320-32EA 176 USD
RC8320-32 RC8320-1 RC8320-5
Sulfuric acid 3.00 N in aqueous solution analytical reagent
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 3.000 ± 0.001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR