Sulfuric acid 2.00 N in aqueous solution

Supplier: Ricca Chemical

8310-16
RC8310-16EA 33 USD
RC8310-16
Sulfuric acid 2.00 N in aqueous solution
Sulfuric acid
Formula: H₂SO₄
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Assay 2 ± 0.001 N at 20 °C
NIST SRM 84


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR