Sulfuric acid 2.00 N in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

R8310000-1A R8310000-4A R8310000-2.5D R8310000-20F
RC831032EA 68 USD
RC831032 RC8310-1 RC8310-5PT RCR8310000-20F
Sulfuric acid 2.00 N in aqueous solution analytical reagent
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8 III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 1.999-2.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR