Sulfuric acid 0.1142 N suitable for Kjeldahl nitrogen analysis of feed grains

Supplier: Ricca Chemical

R8255000-4A R8255000-20F
RC82551EA 185 USD
RC82551 RC82555
Sulfuric acid 0.1142 N suitable for Kjeldahl nitrogen analysis of feed grains
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.1142 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR