Sulfuric acid 0.0400 N (N/25) in aqueous solution analytical reagent

Supplier: Ricca Chemical

R8215000-4A R8215000-1A
RC8215-1EA 124 USD
RC8215-1 RC821532
Sulfuric acid 0.0400 N (N/25) in aqueous solution analytical reagent
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.04000 ± 0.00002 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR