Sulfuric acid 0.0200 N (N/50) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

R8200000-4A R8200000-10F R8200000-1A R8200000-20F R8200000-500A
RC82001EA 137 USD
RC82001 RC820025 RC820032 RC82005 RCR8200000500A
Sulfuric acid 0.0200 N (N/50) in aqueous solution analytical reagent
Sulfuric acid

APHA, for alkalinity. Standardized at 25°C against chemicals whose certification is traceable to NIST.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Tris(hydroxymethyl)aminomethane/Methyl Red) 0.01998-0.02002 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR