Sulfuric acid 0.0200 N (N/50) aqueous solution analytical reagent
Supplier: Ricca Chemical
|
Danger
|
R8200000-4A
R8200000-10F
R8200000-1A
R8200000-20F
R8200000-500A
RC82001EA
137
USD
RC82001
RC820025
RC820032
RC82005
RCR8200000500A
Sulfuric acid 0.0200 N (N/50) aqueous solution analytical reagent
Sulfuric acid
APHA, for alkalinity. Standardized at 25°C against chemicals whose certification is traceable to NIST.
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Tris(hydroxymethyl)aminomethane/Methyl Red) | 0.01998-0.02002 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...