Arsenic(III) oxide standard 0.100 N (N/10) analytical reagent
Supplier: Ricca Chemical
Synonyms:
di-Arsenic trioxide
R0820000-500A
R0820000-4A
RC82016EA
92
USD
RC82016
RC820-1
Arsenic(III) oxide standard 0.100 N (N/10) analytical reagent
Arsenic (III) oxide
Arsenite Standard, RICCA Reagents for Published Test Methods
Contains:
Water ACS/ASTM/USP/EP
Arsenic Trioxide 1327-53-3 ACS
Sodium Hydroxide 1310-73-2 ACS
Sodium Bicarbonate 144-55-8
Formula:
As₂O₃ Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003433 CAS Number: 1327-53-3 EINECS: 231-639-5 UN: 1561 ADR: 6.1,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 83) | 0.1000 ± 0.0005 N at 20.0 °C |
Standard Arsenite Solution, 0.1 N | APHA (4500-Cl C) |
Sodium Arsenite (0.1 N) | EPA (SW-846) (9012A) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...