Sulfuric acid 50% (v/v) in aqueous solution (1 + 1)

Supplier: Ricca Chemical
Danger

R8180000-4A R8180000-500A R8180000-1A R8180000-2.5D R8180000-20F
RC8180-1EA 295 USD
RC8180-1 RC818016 RC818032 RC81805PT RC8180-5
Sulfuric acid 50% (v/v) in aqueous solution (1 + 1)
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.1…1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless to slight yellow, odorless
Assay at 20 °C (traceable to NIST SRM 84) 50.0 ± 0.5 % (v/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR