Sulfuric acid 25% (v/v) in aqueous solution (1 + 3) analytical reagent

Supplier: Ricca Chemical
Danger

R8170000-20F R8170000-1A R8170000-4A R8170000-500A
RC8170-5EA 594 USD
RC8170-5 RC817032 RC8170-1 RC8170-16
Sulfuric acid 25% (v/v) in aqueous solution (1 + 3) analytical reagent
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.1…1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 25.00 ± 0.03 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR