Sulfuric acid 0,7% (v/v) in aqueous solution

Supplier: Ricca Chemical

R8146000-500A R8146000-1A
RC814616EA 39 USD
RC814616 RC8146-32
Sulfuric acid 0,7% (v/v) in aqueous solution
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.700 ± 0.003 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR