Sulfuric acid, dilute R 98 g/L

Supplier: Ricca Chemical
Danger

R8140000-500A
RC8140-16EA 62 USD
RC8140-16
Sulfuric acid, dilute R 98 g/L
Sulfuric acid, dilute R
Formula: H₂O₄S
MW: 98.072 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0400 g/cm³
Storage Temperature: Ambient
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 2.000 ± 0.005 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR