Sulfate Standard Stock Solution, Alcoholic, 1000 ppm SO₄²⁻ in 30% (v/v) Alcohol, Ricca Chemical Company
Supplier: Ricca Chemical
R8111000-500A
RC8111-16EA 159 USD
RC8111-16
Sulfate Standard Stock Solution, Alcoholic, 1000 ppm SO₄²⁻ in 30% (v/v) Alcohol, Ricca Chemical Company
Standards Anionic Standards Sulphate Standards