Starch 2% in aqueous solution stabilized for iodometric titrations mercury-free

Supplier: Ricca Chemical

R8055000-500A R8055000-4A R8055000-20F
RC805516EA 113 USD
RC805516 RC8055-1 RC8055-5
Starch 2% in aqueous solution stabilized for iodometric titrations mercury-free
Starch
Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now

Specification Test Results

Appearance Translucent, odorless
Suitability for Use Colorless (Iodine absent) - Blue (Iodine present)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR