Starch solution R Reag. Ph. Eur. 1085103

Supplier: Ricca Chemical

R8046000-500A
RC8046000-500AEA 166 USD
RC8046000-500A
Starch solution R Reag. Ph. Eur. 1085103
Starch

Used for oxidation-reduction analysis.

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
MDL Number: MFCD00082026
CAS Number: 9005-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR