Starch indicator Platner's

Supplier: Ricca Chemical
Danger

R8001000-500A
RC8001-16EA 133 USD
RC8001-16
Starch indicator Platner's
Starch

White, translucent liquid.

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0500 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8

Order Now

Specification Test Results

Appearance
Suitability for Use Colorless (Iodine absent) - Blue (Iodine present)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR