Sodium thiosulfate 0.0564 N

Supplier: Ricca Chemical
Warning

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

R7941000-4C
RC7941-1EA 380 USD
RC7941-1
Sodium thiosulfate 0.0564 N
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0070 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7

Order Now

Specification Test Results

Appearance Clear, colorless, slight organic odor
Assay at 20 °C (traceable to NIST SRM 136) 0.05640 ± 0.00002 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR