Sodium hydroxide 0,16% (w/v) in aqueous solution for dilution, for cyanide determination

Supplier: Ricca Chemical

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7474000-500A R7474000-1A R7474000-4A
RC747416EA 196 USD
RC747416 RC747432 RC7474-1
Sodium hydroxide 0,16% (w/v) in aqueous solution for dilution, for cyanide determination
Sodium hydroxide
Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.160 ± 0.002 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR