Sodium hydroxide 0.100 N (N/10) in methanol
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
R7472000-4A
R7472000-1A
R7472000-20F
RC7472-1CS
1150
USD
RC7472-1
RCR7472000-1A
RCR7472000-20F
Sodium hydroxide 0.100 N (N/10) in methanol
Sodium hydroxide
Formula:
NaOH Melting Pt: –98°C Density: 0.8 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Phenolphthalein) | 0.0995-0.1005 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...