Sodium hydroxide 6.00 N in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7466000-4A R7466000-1A R7466000-20F R7466000-10F R7466000-500A
RC7466-1EA 220 USD
RC7466-1 RC746632 RC7466-5 RCR7466000-10F RCR7466000500A
Sodium hydroxide 6.00 N in aqueous solution
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: Approximately 0°C
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 6.000 ± 0.005 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR