Sodium hydroxide 5.00 N in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7465000-4A R7465000-1A R7465000-500A R7465000-20F R7465000-10F
RC7465-1EA 192 USD
RC7465-1 RC746532 RC7465-16 RC7465-5 RC7465-2.5
Sodium hydroxide 5.00 N in aqueous solution analytical reagent
Sodium hydroxide
Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 5.000 ± 0.005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR