Sodium hydroxide 2.50 N analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7457000-4A R7457000-1A R7457000-20F R7457000-10F R7457000-500A
RC7457-1EA 139 USD
RC7457-1 RC745732 RC7457-5 RCR7457000-10F RCR7457000500A
Sodium hydroxide 2.50 N analytical reagent
Sodium hydroxide
Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: ∼ 0°C
Density: 1.1
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 2.499-2.501 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR