Sodium hydroxide 1.00 Normal, Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7450000-4F R7450000-10F R7450000-1A R7450000-20F R7450000-4A R7450000-120A R7450000-20E1
RC7450-1CTEA 139 USD
RC7450-1CT RC745025 RC745032 RC74505 RC74501 RC7450-4 RCR745000020E1
Sodium hydroxide 1.00 Normal, Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Sodium hydroxide

Standardized at 25°C against chemicals whose certification is traceable to NIST.

Formula: NaOH
Melting Pt: ∼ 0°C
Density: 1.04
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.999-1.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR