Sodium hydroxide 0.500 Normal (N/2), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical
Warning

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7400000-4A R7400000-10F R7400000-20F R7400000-1A R7400000-120A
RC74001EA 124 USD
RC74001 RC740025 RC74005 RC740032 RC7400-4
Sodium hydroxide 0.500 Normal (N/2), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Sodium hydroxide

Standardized at 25°C against chemicals whose certification is traceable to NIST.

Formula: NaOH
Boiling Pt: ∼105 °C (1013 hPa)
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.5000 ± 0.0002 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR