Sodium hydroxide 0.500 N (N/2) in aqueous solution
Supplier: Ricca Chemical
|
Warning
|
R7400000-4A
R7400000-10F
R7400000-20F
R7400000-1A
R7400000-120A
RC74001EA
124
USD
RC74001
RC740025
RC74005
RC740032
RC7400-4
Sodium hydroxide 0.500 N (N/2) in aqueous solution
Sodium hydroxide
Standardized at 25°C against chemicals whose certification is traceable to NIST.
Formula:
NaOH Boiling Pt: ∼105 °C (1013 hPa) Density: 1.02 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 0.5000 ± 0.0002 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...