Sodium hydroxide 0.250 Normal (N/4), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical
Warning

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7370000-1A R7370000-20F R7370000-4A R7370000-10F R7370000-500A
RC737032EA 56 USD
RC737032 RC73705 RC73701 RC7370-2.5 RCR7370000500A
Sodium hydroxide 0.250 Normal (N/4), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Sodium hydroxide

Clear, colorless liquid.

Formula: NaOH
Boiling Pt: ∼105 °C (1013 hPa)
Melting Pt: 0.0°C
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.2499-0.2501 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR