Sodium hydroxide 0.125 Normal (N/8), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7356000-4A R7356000-20F
RC73561EA 134 USD
RC73561 RC73565
Sodium hydroxide 0.125 Normal (N/8), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Sodium hydroxide
Formula: NaOH
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.1250 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR