Sodium hydroxide 0.100 Normal (N/10), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7350000-500A R7350000-10F R7350000-20F R7350000-4A R7350000-1A
RC735016EA 49 USD
RC735016 RC735025 RC73505 RC73501 RC735032
Sodium hydroxide 0.100 Normal (N/10), Tightrant™ analytical reagent Tested in an ISO 17025 Accredited Facility
Sodium hydroxide

Standardized at 25°C against chemicals whose certification is traceable to NIST.

Formula: NaOH
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.1000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR