Sodium hydroxide 0.0227 Normal (1 ml = 1 mg CO2), Tightrant™ Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7310000-4A R7310000-1A
RC73101EA 122 USD
RC73101 RC731032
Sodium hydroxide 0.0227 Normal (1 ml = 1 mg CO2), Tightrant™ Tested in an ISO 17025 Accredited Facility
Sodium hydroxide

1 mL = 1 mg CO2

Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.02270 ± 0.00005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR