Sodium hydroxide 0.0100 N (N/100) analytical reagent

Supplier: Ricca Chemical

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7295000-4A R7295000-20F R7295000-1A R7295000-10F R7295000-500A
RC72951EA 122 USD
RC72951 RC72955 RC729532 RC7295-2.5 RCR7295000500A
Sodium hydroxide 0.0100 N (N/100) analytical reagent
Sodium hydroxide
Formula: NaOH
Melting Pt: 0.0°C
Density: 1
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 0.00990-0.01010 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR