Sodium hydroxide 40% (w/w) in aqueous solution analytical reagent
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
R7288000-20F
R7288000-1A
R7288000-10F
R7288000-500A
RC7288-5EA
418
USD
RC7288-5
RC7288-32
RC7288-2.5
RCR7288000500A
Sodium hydroxide 40% (w/w) in aqueous solution analytical reagent
Sodium hydroxide
Formula:
NaOH Boiling Pt: ~100 °C Melting Pt: ~0 °C Density: 1.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance | Colorless liquid |
Assay at 20 °C (traceable to NIST SRM 723) | 39.8 - 40.2 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...