Sodium hydroxide 33,3% (w/w) in aqueous solution

Supplier: Ricca Chemical

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

7278.3-1
RC7278.3-1EA 80 USD
RC7278.3-1
Sodium hydroxide 33,3% (w/w) in aqueous solution
Sodium hydroxide
Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Assay 33.3 ± 1 % (w/w)
NIST SRM 723


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR