Sodium hydroxide 30% (w/v) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7274000-500A R7274000-4A R7274000-1A
RC7274-16EA 87 USD
RC7274-16 RC7274-1 RC7274-32
Sodium hydroxide 30% (w/v) in aqueous solution analytical reagent
Sodium hydroxide
Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 7.50 ± 0.05 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR