Sodium hydroxide 10% (w/v) in aqueous solution (100 g/L) analytical reagent
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
R7260000-500A
R7260000-1A
R7260000-120A
R7260000-4A
R7260000-20F
RC726016EA
48
USD
RC726016
RC726032
RC72604
RC7260-1
RCR7260000-20F
Sodium hydroxide 10% (w/v) in aqueous solution (100 g/L) analytical reagent
Sodium hydroxide
Formula:
NaOH Boiling Pt: ~100 °C Melting Pt: ~0 °C Density: 1.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Phenolphthalein) | 9.96 - 10.04 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...