Sodium hydroxide 5% (w/v) in aqueous solution (50 g/L) analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

R7250000-500A R7250000-1A R7250000-4A R7250000-20F
RC725016EA 45 USD
RC725016 RC725032 RC7250-1 RCR7250000-20F
Sodium hydroxide 5% (w/v) in aqueous solution (50 g/L) analytical reagent
Sodium hydroxide
Formula: NaOH
Boiling Pt: ~100 °C
Melting Pt: ~0 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 4.96 - 5.04 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR